Order Entry
Poland
ContactUsLinkComponent
Anti-STAT3 Rabbit Monoclonal Antibody [clone: E121-21]
Anti-STAT3 Rabbit Monoclonal Antibody [clone: E121-21]
Catalog # ORIGTA300380
Supplier:  OriGene
Anti-STAT3 Rabbit Monoclonal Antibody [clone: E121-21]
Catalog # ORIGTA300380
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Signal Transducer And Activator of Transcription 3
  • Antigen symbol:
    STAT3
  • Clonality:
    Monoclonal
  • Clone:
    E121-21
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    PBS 49%,Sodium azide 0,01%,Glycerol 50%,BSA 0,05%
  • Molecular weight:
    88 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µl
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: Western Blot: 1:1000; Immunoprecipitation: 1:20

Type: Primary
Antigen: STAT3 (signal transducer and activator of transcription 3 (acute-phase response factor))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human