Anti-Thy1.2 Rat Monoclonal Antibody (violetFluor® 450) [clone: 30-H12]
Supplier: Tonbo Biosciences
Certificates
About this item
The 30-H12 antibody reacts with mouse CD90.2 (Thy1.2). CD90.2 is a strain-specific allelic form of the GPI-linked membrane associated protein CD90 and is involved in adhesion and signal transduction. CD90.2 is expressed on thymocytes, mature T cells and neurons in mouse strains that express the CD90.2 allele (BALB/c, CBA, C3H, C57BL/6, SJL and others). 30-H12 does not react with the CD90.1 allele expressed in mouse strains such as PL and AKR.
Type: Primary
Antigen: Thy1.2 (Thy1.2)
Clonality: monoclonal
Clone: 30-H12
Conjugation: violetFluor® 450
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Specifications
- Catalog No:
- 75-0903-U025
- 75-0903-U100
- Antigen symbol:
- THY1
- THY1
- Antigen name:
- Thy1 Cell Surface Antigen
- Thy1 Cell Surface Antigen
- Antigen synonyms:
- Thy-1 T-cell antigen|Thy-1 cell surface antigen|Thy-1 membrane glycoprotein|THY1|FLJ33325
- Thy-1 T-cell antigen|Thy-1 cell surface antigen|Thy-1 membrane glycoprotein|THY1|FLJ33325
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- VioletFluor® 450
- VioletFluor® 450
- Clone:
- 30-H12
- 30-H12
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 21838
- 21838
- Isotype:
- IgG2b
- IgG2b
- Flow cytometry:
- Yes
- Yes
- Cross adsorption:
- none
- none
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together