Anti-CD1E Rabbit Polyclonal Antibody [clone: 406CT21.4.3]
ABGEAP9942C
: Abgent
- Antibody type:Primary
- Antigen name:CD1E molecule
- Antigen symbol:CD1E
- Clonality:Polyclonal
- Clone:406CT21.4.3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgM
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Form:Ascites
- Gene ID:796
- Antigen synonyms:CD1e|hCD1e|sCD1e|T-cell surface glycoprotein CD1e|soluble|R2G1|membrane-associated
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:KRDMSSDLERDHRPC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Shipping temperature:4 °C
- Immunogen:This CALCA/CT antibody is generated from mice immunized with a KLH conjugated synthetic peptide between 104-132 amino acids from human CALCA/CT.
- Pk:400 µl
Type: Primary
Antigen: CD1E (CD1e molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human