Order Entry
Poland
ContactUsLinkComponent
Anti-ZRANB1 Rabbit Polyclonal Antibody
Anti-ZRANB1 Rabbit Polyclonal Antibody
Catalog # ABGEAP9830B
Supplier:  Abgent
undefined
Anti-ZRANB1 Rabbit Polyclonal Antibody
Catalog # ABGEAP9830B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Zinc Finger RAN-binding Domain Containing 1
  • Antigen symbol:
    ZRANB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-Terminal
  • Gene ID:
    30009
  • Antigen synonyms:
    T-box transcription factor TBX21|TBX21 |T-cell-specific T-box transcription factor T-bet|T-PET|T-box protein 21|T-bet|TBLYM|TBET
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Sequence:
    YPEPGAQDADERRGC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This TBX21 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 22-50 amino acids from the N-terminal region of human TBX21.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: ZRANB1 (zinc finger, RAN-binding domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse