Order Entry
Poland
ContactUsLinkComponent
Anti-TLR4 Rabbit Polyclonal Antibody
Anti-TLR4 Rabbit Polyclonal Antibody
  ABGEAP9685C
 :  Abgent
undefined
Anti-TLR4 Rabbit Polyclonal Antibody
  ABGEAP9685C
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Toll-like Receptor 4
  • Antigen symbol:
    TLR4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-Terminal
  • Gene ID:
    4343
  • Antigen synonyms:
    TLR4|Toll-like receptor 4|Rasl2-8|TLR-4|Lps|Ran/M1|Ly87|hToll|CD284|ARMD10|TOLL
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Sequence:
    ERPDRAKGYDLELSC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0,4 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This MOV10 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 260-288 amino acids from the N-terminal region of human MOV10.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: TLR4 (toll-like receptor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human