Anti-COXI Rabbit Polyclonal Antibody
Catalog # ABGEAP9684C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Cytochrome c oxidase subunit 1
- Antigen symbol:COX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:Center
- Gene ID:57332
- Antigen synonyms:PCOX1|PGHS1|PHS1|COX3|COX1|PES-1|PGHS-1|PGG/HS|PTGHS
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:RERERGTSRVDDKPC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,35 mg/ml
- Shipping temperature:4 °C
- Immunogen:This CBX8 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 161-189 amino acids from the Central region of human CBX8.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: COXI
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human