Anti-IRS2 Rabbit Polyclonal Antibody
Catalog # ABGEAP7789B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Insulin Receptor Substrate 2
- Antigen symbol:IRS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: IRS2 (insulin receptor substrate 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human