Anti-YARS Rabbit Polyclonal Antibody
ABGEAP7580B
: Abgent
- Antibody type:Primary
- Antigen name:Tyrosyl-tRNA Synthetase 1, Cytoplasmic
- Antigen symbol:YARS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:CMTDIC|YTS|YRS|tyrosyl-tRNA synthetase|TYRRS
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: YARS (tyrosyl-tRNA synthetase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human