Anti-CAMK1D Rabbit Polyclonal Antibody
ABGEAP7204B
: Abgent
- Antibody type:Primary
- Antigen name:Calcium/calmodulin-dependent protein kinase type 1D
- Antigen symbol:CAMK1D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:CaM-KI delta|CaM kinase I delta|CKLiK|CamKI-like protein kinase|Camk1D|CaM kinase ID|CaMKI delta|CaMKI-delta|CAMK1D
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: CAMK1D (calcium/calmodulin-dependent protein kinase ID)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human