Anti-LMTK2 Rabbit Polyclonal Antibody
Catalog # ABGEAP7140C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Lemur tyrosine kinase 2
- Antigen symbol:LMTK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:LMR2|KPI-2|BREK|AATYK2|cprk|PPP1R100|KPI2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: LMTK2 (lemur tyrosine kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human