Anti-CAMKK2 Rabbit Polyclonal Antibody
Catalog # ABGEAP7117B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Calcium/Calmodulin-dependent Protein Kinase 2 Beta
- Antigen symbol:CAMK2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:Calcium/calmodulin-dependent protein kinase kinase 2|KIAA0787|CaM-kinase kinase beta|CaM-KK beta|Calcium/calmodulin-dependent protein kinase kinase beta|CAMKK2|CaMKK beta|CAMKKB
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: CAMKK2 (calcium/calmodulin-dependent protein kinase kinase 2, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human