Anti-C6 Rabbit Polyclonal Antibody
Catalog # ABGEAP6937A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Complement Component 6
- Antigen symbol:C6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:CP24|HCAI|CYP24|P450-CC24
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: C6 (complement component 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human