Anti-IPO9 Rabbit Polyclonal Antibody
Catalog # ABGEAP5564A
Dostawca: Abgent
Dane techniczne
- Antibody type:Primary
- Antigen name:Importin 9
- Antigen symbol:IPO9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Dane techniczne
Informacje o tym produkcie
Type: Primary
Antigen: IPO9 (importin 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse