Order Entry
Poland
ContactUsLinkComponent
 
Anti-ADH1C Rabbit Polyclonal Antibody
Catalog # ABGEAP5534C
Supplier:  Abgent
undefined
Anti-ADH1C Rabbit Polyclonal Antibody
Catalog # ABGEAP5534C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Alcohol Dehydrogenase 1C
  • Antigen symbol:
    ADH1C
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    middle
  • Antigen synonyms:
    ADH3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: ADH1C (alcohol dehydrogenase 1C (class I), gamma polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human