Anti-DHRS7 Rabbit Polyclonal Antibody
ABGEAP4772A
: Abgent
- Antibody type:Primary
- Antigen name:Dehydrogenase/reductase (SDR family) Member 7
- Antigen symbol:DHRS7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:DHRS7_HUMAN.|CGI86|Retinal short chain dehydrogenase/reductase 4|RETSDR4|DHRS7A|Dehydrogenase/reductase SDR family member 7|CGI 86|retDSR4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: DHRS7 (dehydrogenase/reductase (SDR family) member 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human