Anti-MDM2 Rabbit Polyclonal Antibody
ABGEAP3490A
: Abgent
- Antibody type:Primary
- Antigen name:Mouse Double Minute 2
- Antigen symbol:MDM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:1700007J15Rik|Double minute 2 protein|AA415488|HDMX|Ubiquitin-protein ligase E3 Mdm2|Mdm-2|ACTFS|Oncoprotein Mdm2|p53-binding protein Mdm2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: MDM2 (MDM2 oncogene, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human