Anti-RFC5 Rabbit Polyclonal Antibody
Catalog # ABGEAP2812C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Replication factor C subunit 5
- Antigen symbol:RFC5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:Activator 1 subunit 5|Replication factor C subunit 5|RF-C 36 kDa subunit|RFC5|Replication factor C 36 kDa subunit|A1 36 kDa subunit|RFC36|Activator 1 36 kDa subunit
- Sequence:R185
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: RFC5 (replication factor C (activator 1) 5, 36.5kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Rat