Order Entry
Poland
ContactUsLinkComponent
Anti-UBQLN3 Rabbit Polyclonal Antibody
Anti-UBQLN3 Rabbit Polyclonal Antibody
Catalog # ABGEAP2178B
Supplier:  Abgent
undefined
Anti-UBQLN3 Rabbit Polyclonal Antibody
Catalog # ABGEAP2178B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ubiquilin 3
  • Antigen symbol:
    UBQLN3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal ( 587–601 aa)
  • Cross adsorption:
    No
  • Antigen synonyms:
    SHP2|PTP2C|PTP-1D|SH-PTP3|METCDS|NS1|CFC|SH-PTP2|BPTP3|JMML
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: UBQLN3 (ubiquilin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse