Order Entry
Poland
ContactUsLinkComponent
Anti-PYCARD Rabbit Polyclonal Antibody
Anti-PYCARD Rabbit Polyclonal Antibody
Catalog # ABGEAP13862B
Dostawca:  Abgent
Anti-PYCARD Rabbit Polyclonal Antibody
Catalog # ABGEAP13862B
Dostawca:  Abgent

Dane techniczne

  • Antibody type:
    Primary
  • Antigen name:
    PYD and CARD domain containing
  • Antigen symbol:
    PYCARD
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Antigen synonyms:
    ASC, ASC, TMS, TMS1, CARD5, TMS-1, ASC, Apoptosis-associated speck-like protein containing a CARD, Caspase recruitment domain-containing protein 5, hASC
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Dane techniczne

Informacje o tym produkcie

Type: Primary
Antigen: PYCARD (PYD and CARD domain containing)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human