Anti-IL21 Rabbit Polyclonal Antibody
Catalog # ABGEAP13840B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 21
- Antigen symbol:IL21
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Antigen synonyms:Za11|Interleukin-21|IL-21
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: IL21 (interleukin 21)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human