Anti-ZNF222 Rabbit Polyclonal Antibody
Catalog # ABGEAP13483A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger Protein 222
- Antigen symbol:ZNF222
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:EWS-WT1|WAGR|WT33|AWT1|NPHS4|WIT-2|GUD
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: ZNF222 (zinc finger protein 222)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human