Anti-ABHD2 Rabbit Polyclonal Antibody
Catalog # ABGEAP13083C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Abhydrolase Domain Containing 2
- Antigen symbol:ABHD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:LABH2|HS1-2|PHPS1-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: ABHD2 (abhydrolase domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human