Anti-SCHIP1 Rabbit Polyclonal Antibody
numer katalogowy ABGEAP12855C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Schwannomin interacting protein 1
- Antigen symbol:SCHIP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:carnitine acyltransferase-like protein 1|FLJ23809|CPTIC|CATL1|CPT1P|CPT1-like pseudogene|carnitine palmitoyltransferase 1C|CPT1C|carnitine palmitoyltransferase I related C
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: SCHIP1 (schwannomin interacting protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human