Order Entry
Poland
ContactUsLinkComponent
Anti-CTNND2 Rabbit Polyclonal Antibody
Anti-CTNND2 Rabbit Polyclonal Antibody
numer katalogowy ABGEAP12548B
Supplier:  Abgent
Anti-CTNND2 Rabbit Polyclonal Antibody
numer katalogowy ABGEAP12548B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Catenin Delta 2
  • Antigen symbol:
    CTNND2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Antigen synonyms:
    GT24|CTND2_HUMAN|Neural plakophilin related arm repeat protein|Delta catenin|Catenin Cadherin associated protein delta 2|CTNND2|Neurojungin|Catenin delta 2|NPRAP|CTNND 2|Catenin delta-2|Neural plakophilin related armadillo repeat protein|Delta-catenin|GT 24|Neural plakophilin-related ARM-repeat protein
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: CTNND2 (catenin (cadherin-associated protein), delta 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse