Order Entry
Poland
ContactUsLinkComponent
Anti-MDM2 Rabbit Polyclonal Antibody
Anti-MDM2 Rabbit Polyclonal Antibody
Catalog # ABGEAP1253E
Supplier:  Abgent
undefined
Anti-MDM2 Rabbit Polyclonal Antibody
Catalog # ABGEAP1253E
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mouse Double Minute 2
  • Antigen symbol:
    MDM2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Antigen synonyms:
    1700007J15Rik|Double minute 2 protein|AA415488|HDMX|Ubiquitin-protein ligase E3 Mdm2|Mdm-2|ACTFS|Oncoprotein Mdm2|p53-binding protein Mdm2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: MDM2 (MDM2 oncogene, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human