Anti-OTOP3 Rabbit Polyclonal Antibody
ABGEAP11912B
: Abgent
- Antibody type:Primary
- Antigen name:Otopetrin 3
- Antigen symbol:OTOP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: OTOP3 (otopetrin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human