Anti-IRGM Rabbit Polyclonal Antibody
Catalog # ABGEAP11128B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen symbol:IRGM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Antigen synonyms:IP3 3-kinase A|Inositol-trisphosphate 3-kinase A|Inositol 1,4,5-trisphosphate 3-kinase A|IP3K A
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: IRGM (immunity-related GTPase family, M)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human