Order Entry
Poland
ContactUsLinkComponent
Anti-CITED2 Rabbit Polyclonal Antibody
Anti-CITED2 Rabbit Polyclonal Antibody
Catalog # ABGEAP10174B
Supplier:  Abgent
Anti-CITED2 Rabbit Polyclonal Antibody
Catalog # ABGEAP10174B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal Domain, 2
  • Antigen symbol:
    CITED2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal ( 600–650 aa)
  • Cross adsorption:
    No
  • Antigen synonyms:
    P35SRJ|MRG1|Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: CITED2 (Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse