Anti-PAX6 Mouse Monoclonal Antibody
ABGEAM1871B
: Abgent
- Antibody type:Primary
- Antigen name:Paired box 6
- Antigen symbol:PAX6
- Clonality:Monoclonal
- Clone:193CT15.2.2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:1–100 aa
- Cross adsorption:No
- Antigen synonyms:MGDA|Paired Box Protein-6|Oculorhombin|WAGR|Keratitis)|Keratitis|AN2|Aniridia|Aniridia Type II Protein|Paired Box Gene 6 (Aniridia
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: PAX6 (paired box 6)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human