Anti-PAG1 Rabbit Polyclonal Antibody
Catalog # AVIVARP63792_P050
Dostawca: Aviva Systems Biology
Dane techniczne
- Antibody type:Primary
- Antigen name:phosphoprotein membrane anchor with glycosphingolipid microdomains 1
- Antigen symbol:PAG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:55824
- Antigen synonyms:CBP|PAG
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Dane techniczne
Informacje o tym produkcie
Type: Primary
Antigen: PAG1 (phosphoprotein associated with glycosphingolipid microdomains 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: