Anti-GPR19 Rabbit Polyclonal Antibody
AVIVARP62972_P050
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor 19
- Antigen symbol:GPR19
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:2842
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: GPR19 (G protein-coupled receptor 19)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: