Anti-MTMR14 Rabbit Polyclonal Antibody
Catalog # AVIVARP57637_P050
Dostawca: Aviva Systems Biology
Dane techniczne
- Antibody type:Primary
- Antigen name:Myotubularin Related Protein 14
- Antigen symbol:MTMR14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:64419
- Antigen synonyms:C3orf29
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Dane techniczne
Informacje o tym produkcie
Type: Primary
Antigen: MTMR14 (myotubularin related protein 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: