Anti-RANGAP1 Rabbit Polyclonal Antibody
AVIVARP56510_P050
- Antibody type:Primary
- Antigen name:Ran GTPase activating protein 1
- Antigen symbol:RANGAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:19387
- Antigen synonyms:C79654|Fug1|mKIAA1835
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: RANGAP1 (Ran GTPase activating protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: