Anti-KIAA1191 Rabbit Polyclonal Antibody
AVIVARP56289_P050
- Antibody type:Primary
- Antigen symbol:KIAA1191
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:57179
- Antigen synonyms:p60MONOX
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: KIAA1191 (KIAA1191)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: