Anti-GPX7 Rabbit Polyclonal Antibody
Catalog # AVIVARP55329_P050
Dostawca: Aviva Systems Biology
Dane techniczne
- Antibody type:Primary
- Antigen name:Glutathione peroxidase 7
- Antigen symbol:GPX7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:298376
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Dane techniczne
Informacje o tym produkcie
Type: Primary
Antigen: GPX7 (glutathione peroxidase 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Rat