Anti-APOE Rabbit Polyclonal Antibody
AVIVARP54283_P050
- Antibody type:Primary
- Antigen name:apolipoprotein E
- Antigen symbol:APOE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:348
- Antigen synonyms:APO-E|AD2|LDLCQ5|LPG
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: APOE (apolipoprotein E)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human