Anti-APOA1 Rabbit Polyclonal Antibody
AVIVARP54279_P050
- Antibody type:Primary
- Antigen name:Apolipoprotein A1
- Antigen symbol:APOA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:25081
- Antigen synonyms:apoA-I
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: APOA1 (apolipoprotein A-I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: