Anti-SQSTM1 Rabbit Polyclonal Antibody
Catalog # AVIVARP54262_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:sequestosome 1
- Antigen symbol:SQSTM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:8878
- Antigen synonyms:OSIL|A170|FTDALS3|ZIP3|p60|p62B|p62|PDB3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SQSTM1 (sequestosome 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: