Anti-PPP3CA Rabbit Polyclonal Antibody
AVIVARP51337_P050
- Antibody type:Primary
- Antigen name:protein phosphatase 3, catalytic subunit, alpha isozyme
- Antigen symbol:PPP3CA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:5530
- Antigen synonyms:CALN|CCN1|PPP2B|CNA1|CALNA1|CALNA
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: PPP3CA (protein phosphatase 3, catalytic subunit, alpha isozyme)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: