Anti-DLK2 Rabbit Polyclonal Antibody
Catalog # AVIVARP49784_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:delta-like 2 homolog (Drosophila)
- Antigen symbol:DLK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:106565
- Antigen synonyms:DLK-2|Egfl9|AI413481
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: DLK2 (delta-like 2 homolog (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: