Order Entry
Poland
ContactUsLinkComponent
Anti-KIAA1324 Rabbit Polyclonal Antibody
Anti-KIAA1324 Rabbit Polyclonal Antibody
  AVIVARP49563_P050
Anti-KIAA1324 Rabbit Polyclonal Antibody
  AVIVARP49563_P050

 

  • Antibody type:
    Primary
  • Antigen symbol:
    KIAA1324
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    57535
  • Antigen synonyms:
    EIG121
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: KIAA1324 (KIAA1324)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: