Anti-FMO3 Rabbit Polyclonal Antibody
Catalog # AVIVARP44433_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Flavin containing monooxygenase 3
- Antigen symbol:FMO3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:2328
- Antigen synonyms:dJ127D3.1|FMOII|TMAU
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: FMO3 (flavin containing monooxygenase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: