Anti-MTUS1 Rabbit Polyclonal Antibody
Catalog # AVIVARP44418_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:microtubule associated tumor suppressor 1
- Antigen symbol:MTUS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:57509
- Antigen synonyms:ATBP|ATIP|MP44|ICIS|MTSG1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: MTUS1 (microtubule associated tumor suppressor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: