Peqlab PerfectBlue™ Twin L, Dual Gel System
Supplier: VWR Peqlab
Certificates
About this item
The largest of the series, the Twin L can accommodate 20×20 cm gels for the simultaneous separation of up to 50 samples making this system ideally suited for either higher throughput or separating lower molecular weight samples. The system actively manages temperature build up with an efficient water cooling system and central aluminium ceramic plates for effective and safe long term operation.
*Based on an average gel and a tooth at 20 mm depth
Ordering information: Complete system, with or without casting base, includes internal cooling core, SuperSafe cover with attached leads, 4× glass plate pair sets, 4× combs (1,5 mm thick, 2× 15 teeth and 2× 20 teeth), 4× spacer sets (1,5 mm), 2× replacement gaskets, 1× blocking plate (for single gel runs), and 1× SpacerPlate.
Specifications
- Catalog No:
- 700-0441
- 700-0658
- Buffer volume, gel tank:
- ~1250 ml
- ~1250 ml
- Sample capacity:
- 10 - 50
- 10 - 50
- Unit W×H×D:
- 300×170×240 mm
- 300×170×240 mm
Specifications
Related Information
Broschüre_PerfectBlue_DE Doppelgelsystem Twin L
Product Literature
PerfectBlue Twin_EN+DE_m
Manual
Frequently Bought Together