Order Entry
Poland
ContactUsLinkComponent
Multipurpose trays, with lid, SP Bel-Art
Multipurpose trays, with lid, SP Bel-Art
Catalog # BELAF162300000
undefined
Multipurpose trays, with lid, SP Bel-Art
Catalog # BELAF162300000
Specifications
  • W×D×H:
    203×273×76 mm
  • Pk:
    1
Specifications
About this item
PP.
  • Chemical and corrosion resistant
  • Flexible and strong
  • Stackable to conserve space
Multipurpose trays with snap on lid is ideal for safely storing small items and holding instruments while autoclaving. Rounded and smooth inside corners allow easy cleaning.