Order Entry
Poland
Orders LinkContactUsLinkComponent
Molecular models
Molecular models
Molecular structure box
Catalog # WILE3-527-32662-4
Supplier:   WILEY JOHN SONS
Molecular models
Catalog # WILE3-527-32662-4
Supplier:  WILEY JOHN SONS
Molecular models
Molecular models
Molecular structure box
Price
Inquire For Price
Log in for availability
1 * 1 SZT
Product Details & Documents
This set contains more than 240 atomic centers (annex C, H, O, N and P, S metals and halogens) and over 200 connections allow the assembly of even complex molecules from all fields of chemistry.
It has also 224 compounds in different lengths.
Ordering information: Includes a colour instruction booklet.
Specifications
  • Description:
    Molecular structure box