0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
Orders Link
ContactUsLinkComponent
Calcium hypochlorite
Calcium hypochlorite 60% available chlorine, TECHNICAL
Calcium hypochlorite 60% available chlorine, TECHNICAL
Calcium hypochlorite 60% available chlorine, TECHNICAL
Calcium hypochlorite 60% available chlorine, TECHNICAL
:
:
7778-54-3
Expand image
Calcium hypochlorite 60% available chlorine, TECHNICAL
Add to cart
:
7778-54-3
:
7778-54-3
:
CaCl₂O₂
:
MFCD00010900
:
142.98 g/mol
:
231-908-7
:
1748
:
5.1,II
:
01-2119487005-40
Documents
Packaging Information
1L Plastic bottle for solids DIN 60 UN (ZPACK13251000)
6L Plastic bottle for solids DIN 80 (ZPACK13256000)
SDS
sds_VWRC22669
Product specification
spec_VWRC22669.293
spec_VWRC22669.293
spec_VWRC22669.293
spec_VWRC22669.362
spec_VWRC22669.362
Assay (Active chlorine)(at filling):
Min. 61 %