



Aniline ≥99.0%, BAKER ANALYZED®, J.T.Baker®
9110-01
: Avantor
:
- Pack type:Glass bottle, amber, narrow mouth
- Pk:500 mL
- Seq:2725
- For Laboratory,Research,or Manufacturing Use:= 99.0 %
- NOTE: This product darkens to a reddish-brown colo r on storage.:= 250
- Assay (C₆H₅NH₂) (by GC, corrected for water):≥99.0 %
- Color (APHA) (at time of packaging):≤250
- Residue after Ignition:≤0.005 %
- Chlorobenzene (C₆H₅Cl):≤0.01 %
- Hydrocarbons:Passes Test
- Nitrobenzene (C₆H₅NO₂):Passes Test
:
:93.13 g/mol
:13,00661
:184 °C (1013 hPa)
:–6.2 °C
:1.019 g/cm³ (20 °C)
:76 °C
:Ambient
:200-539-3
:1547
:6.1,II
:GHS06
:GHS05
:GHS08
:GHS09
:62-53-3
:C₆H₅NH₂
:MFCD00007629