Order Entry
Poland
Orders LinkContactUsLinkComponent
Software, IRsolution Agent
Software, IRsolution Agent
IR solution agent software version 2.2 (single user license)
Catalog # SHIM206-21600-92
Supplier:   SHIMADZU
Software, IRsolution Agent
Catalog # SHIM206-21600-92
Supplier:  SHIMADZU
Software, IRsolution Agent
IR solution agent software version 2.2 (single user license)
Price
4 290 zł
Log in for availability
1 * 10 SZT
Product Details & Documents
IRsolution Agent is a tool for managing data from IRsolution results in a database. This allows managing analytical data records over their entire life cycle, from creation (measurement), review, approval, storage, viewing, backup, and disposal.
  • Provides security management and electronic signature functinos for data stored and recorded in a database
  • Measurement data is automatically saved in a database to allow easy searching of target data at a later time
  • Spectral waveform data, peak detection tables and other information stored in the database can be viewed directly
Certifications: Compliant with FDA 21 CFR Part 11 (electronic records and electronic signatures).
Specifications
  • Description:
    IR solution agent software version 2.2 (single user license)