Order Entry
Poland
ContactUsLinkComponent
 
Screw cap for 22 mm vials
Catalog # HEWL8010-0565
Supplier:  Agilent
Screw cap for 22 mm vials
Catalog # HEWL8010-0565
Supplier:  Agilent

Specifications

  • For:
    10 ml vials
  • Septum:
    -
  • Description:
    Screw cap, black, without septa
  • Pk:
    100

Specifications

About this item